Assay Details
| Gene Symbol: MTHFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129979.3 | NP_566011.1 | 8 - 9 | 1483 | 115 | |
| GenBank mRNA | AF181967.1 | - | 8 - 9 | 1371 | 115 | |
| AY050434.1 | - | 8 - 9 | 1411 | 115 | ||
More Information
Set Membership: |
|

MTHFR2
Genomic Map