Assay Details
Gene Symbol: GL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344395.1 | NP_001332706.1 | 3 - 4 | 871 | 80 | |
NM_001344396.1 | NP_001332705.1 | 4 - 5 | 727 | 80 | ||
NM_148067.4 | NP_680372.1 | 3 - 4 | 506 | 80 | ||
GenBank mRNA | AF246291.1 | - | 3 - 4 | 504 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
