Assay Details
Gene Symbol: GL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344395.1 | NP_001332706.1 | 5 - 6 | 1766 | 107 | |
NM_001344396.1 | NP_001332705.1 | 6 - 7 | 1622 | 107 | ||
NM_148067.4 | NP_680372.1 | 5 - 6 | 1401 | 107 | ||
GenBank mRNA | AF246291.1 | - | 5 - 6 | 1399 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
