Assay Details
| Gene Symbol: ATBET12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179053.3 | NP_849384.1 | 2 - 3 | 333 | 110 | |
| GenBank mRNA | AK117428.1 | - | 2 - 3 | 312 | 110 | |
| BT026096.1 | - | 2 - 3 | 170 | 110 | ||
More Information
Set Membership: |
|

ATBET12
Genomic Map