Assay Details
| Gene Symbol: ATBET12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179053.3 | NP_849384.1 | 4 - 5 | 443 | 109 | |
| GenBank mRNA | AK117428.1 | - | 4 - 5 | 422 | 109 | |
| BT026096.1 | - | 4 - 5 | 280 | 109 | ||
More Information
Set Membership: |
|

ATBET12
Genomic Map