Assay Details
| Gene Symbol: AT2G39690 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336785.1 | NP_001318384.1 | 1 - 2 | 130 | 98 | |
| NM_001336788.1 | NP_001324848.1 | 1 - 2 | 175 | 98 | ||
| NM_001336789.1 | NP_001324850.1 | 1 - 2 | 130 | 98 | ||
| NM_001336790.1 | NP_001324846.1 | 1 - 2 | 130 | 98 | ||
| Gene Symbol: AT3G47675 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339330.1 | NP_001319702.1 | 2 - 3 | 331 | 98 | |
| GenBank mRNA | AY099830.1 | - | 5 - 6 | 802 | 98 | |
| BT000320.1 | - | 1 - 2 | 80 | 98 | ||
More Information
Set Membership: |
|

AT2G39690
Genomic Map