Assay Details
| Gene Symbol: ARPC1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202716.1 | NP_001189645.1 | 15 - 16 | 1388 | 142 | |
| NM_128643.4 | NP_180648.1 | 14 - 15 | 1345 | 142 | ||
| NM_201841.1 | NP_973570.1 | 13 - 14 | 1304 | 142 | ||
| GenBank mRNA | AK227543.1 | - | 14 - 15 | 1304 | 142 | |
More Information
Set Membership: |
|

ARPC1A
Genomic Map