Assay Details
| Gene Symbol: SAR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036392.1 | NP_001031469.1 | 3 - 4 | 686 | 121 | |
| NM_128872.3 | NP_180871.1 | 3 - 4 | 814 | 121 | ||
| GenBank mRNA | AF419564.1 | - | 3 - 4 | 594 | 121 | |
| AK317447.1 | - | 3 - 4 | 621 | 121 | ||
| AY072422.1 | - | 3 - 4 | 597 | 121 | ||
| AY079037.1 | - | 3 - 4 | 456 | 121 | ||
| AY086363.1 | - | 3 - 4 | 662 | 121 | ||
| AY114706.1 | - | 3 - 4 | 456 | 121 | ||
| AY128288.1 | - | 3 - 4 | 594 | 121 | ||
More Information
Set Membership: |
|

SAR1
Genomic Map