Assay Details
| Gene Symbol: AT1G16230 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332215.1 | NP_001319019.1 | 3 - 4 | 471 | 141 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G16230| Gene Symbol: AT1G16230 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332215.1 | NP_001319019.1 | 3 - 4 | 471 | 141 | |
Set Membership: |
|