Assay Details
| Gene Symbol: HMGB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035998.1 | NP_001031075.1 | 4 - 5 | 378 | 79 | |
| NM_001084101.2 | NP_001077570.2 | 1 - 2 | 306 | 79 | ||
| NM_001332458.1 | NP_001322267.1 | 3 - 4 | 831 | 79 | ||
| NM_001332459.1 | NP_001322266.1 | 4 - 5 | 515 | 79 | ||
| NM_101921.4 | NP_564124.1 | 4 - 5 | 546 | 79 | ||
| GenBank mRNA | AF411789.1 | - | 4 - 5 | 262 | 79 | |
| AY084848.1 | - | 4 - 5 | 353 | 79 | ||
| AY097385.1 | - | 3 - 4 | 197 | 79 | ||
| AY136309.1 | - | 4 - 5 | 317 | 79 | ||
| BT000420.1 | - | 3 - 4 | 197 | 79 | ||
| Y09482.1 | - | 245 | 79 | |||
| Y14073.1 | - | 247 | 79 | |||
More Information
Set Membership: |
|

HMGB3
Genomic Map