Assay Details
| Gene Symbol: HMGB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035998.1 | NP_001031075.1 | 5 - 6 | 423 | 134 | |
| NM_001084101.2 | NP_001077570.2 | 2 - 3 | 351 | 134 | ||
| NM_001332458.1 | NP_001322267.1 | 4 - 5 | 876 | 134 | ||
| NM_001332459.1 | NP_001322266.1 | 5 - 6 | 560 | 134 | ||
| NM_101921.4 | NP_564124.1 | 5 - 6 | 591 | 134 | ||
| GenBank mRNA | AF411789.1 | - | 5 - 6 | 307 | 134 | |
| AY084848.1 | - | 5 - 6 | 398 | 134 | ||
| AY097385.1 | - | 4 - 5 | 242 | 134 | ||
| AY136309.1 | - | 5 - 6 | 362 | 134 | ||
| BT000420.1 | - | 4 - 5 | 242 | 134 | ||
| Y09482.1 | - | 290 | 134 | |||
| Y14073.1 | - | 292 | 134 | |||
More Information
Set Membership: |
|

HMGB3
Genomic Map