Assay Details
| Gene Symbol: HMGB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035998.1 | NP_001031075.1 | 7 - 8 | 591 | 121 | |
| NM_001332458.1 | NP_001322267.1 | 6 - 7 | 1045 | 121 | ||
| NM_001332459.1 | NP_001322266.1 | 6 - 7 | 957 | 121 | ||
| NM_101921.4 | NP_564124.1 | 7 - 8 | 760 | 121 | ||
| GenBank mRNA | AF411789.1 | - | 7 - 8 | 476 | 121 | |
| AY084848.1 | - | 7 - 8 | 567 | 121 | ||
| AY136309.1 | - | 7 - 8 | 531 | 121 | ||
| BT000420.1 | - | 6 - 7 | 411 | 121 | ||
| Y09482.1 | - | 459 | 121 | |||
| Y14073.1 | - | 461 | 121 | |||
More Information
Set Membership: |
|

HMGB3
Genomic Map