Assay Details
Gene Symbol: UGT85A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102088.3 | NP_173655.2 | 1 - 2 | 618 | 114 | |
GenBank mRNA | KJ138991.1 | - | 1 - 2 | 523 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
