Assay Details
Gene Symbol: smB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179083.3 | NP_849414.1 | 1 - 2 | 1010 | 142 | |
GenBank mRNA | BT004219.1 | - | 1 - 2 | 907 | 142 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
