Assay Details
Gene Symbol: BIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344430.1 | NP_001331824.1 | 2 - 3 | 641 | 95 | |
NM_123567.3 | NP_199017.2 | 2 - 3 | 431 | 95 | ||
NM_180788.3 | NP_851119.1 | 2 - 3 | 537 | 95 | ||
GenBank mRNA | BT002392.1 | - | 2 - 3 | 431 | 95 | |
BT008406.1 | - | 2 - 3 | 324 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
