Assay Details
Gene Symbol: UBC30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_180867.3 | NP_851198.1 | 4 - 5 | 550 | 137 | |
GenBank mRNA | AY059806.1 | - | 4 - 5 | 377 | 137 | |
AY081511.1 | - | 3 - 4 | 298 | 137 | ||
AY084220.1 | - | 4 - 5 | 383 | 137 | ||
DQ027043.1 | - | 3 - 4 | 298 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
