Assay Details
| Gene Symbol: SYNC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105766.3 | NP_177254.1 | 4 - 5 | 1834 | 128 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SYNC3| Gene Symbol: SYNC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105766.3 | NP_177254.1 | 4 - 5 | 1834 | 128 | |
Set Membership: |
|