Assay Details
Gene Symbol: ARAC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106234.3 | NP_177712.1 | 3 - 4 | 614 | 128 | |
GenBank mRNA | AY062800.1 | - | 3 - 4 | 556 | 128 | |
AY081600.1 | - | 3 - 4 | 299 | 128 | ||
U52350.1 | - | 3 - 4 | 494 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
