Assay Details
Gene Symbol: ARAC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106234.3 | NP_177712.1 | 4 - 5 | 678 | 114 | |
GenBank mRNA | AY062800.1 | - | 4 - 5 | 620 | 114 | |
AY081600.1 | - | 4 - 5 | 363 | 114 | ||
U52350.1 | - | 4 - 5 | 558 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
