Assay Details
| Gene Symbol: LSM3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106337.2 | NP_177812.1 | 2 - 3 | 324 | 168 | |
| GenBank mRNA | AY086266.1 | - | 2 - 3 | 325 | 168 | |
More Information
Set Membership: |
|

LSM3B
Genomic Map