Assay Details
| Gene Symbol: AT1G60680 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333909.1 | NP_001323339.1 | 1 - 2 | 287 | 144 | |
| NM_104751.5 | NP_176267.3 | 1 - 2 | 307 | 144 | ||
| GenBank mRNA | AK227526.1 | - | 1 - 2 | 238 | 144 | |
| BT006462.1 | - | 1 - 2 | 226 | 144 | ||
More Information
Set Membership: | 
		   
					
						
							 | 
				

AT1G60680
Genomic Map