Assay Details
| Gene Symbol: RPL18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337591.1 | NP_001327673.1 | 2 - 3 | 608 | 150 | |
| NM_001337592.1 | NP_001327672.1 | 3 - 4 | 486 | 150 | ||
| NM_111432.4 | NP_187210.1 | 3 - 4 | 380 | 150 | ||
| GenBank mRNA | AY037224.1 | - | 3 - 4 | 255 | 150 | |
| AY060533.1 | - | 2 - 3 | 224 | 150 | ||
More Information
Set Membership: |
|

RPL18
Genomic Map