Assay Details
| Gene Symbol: RPL18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337591.1 | NP_001327673.1 | 3 - 4 | 788 | 172 | |
| NM_001337592.1 | NP_001327672.1 | 4 - 5 | 666 | 172 | ||
| NM_111432.4 | NP_187210.1 | 4 - 5 | 560 | 172 | ||
| GenBank mRNA | AY037224.1 | - | 4 - 5 | 435 | 172 | |
| AY060533.1 | - | 3 - 4 | 404 | 172 | ||
More Information
Set Membership: |
|

RPL18
Genomic Map