Assay Details
| Gene Symbol: MEE67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337855.1 | NP_001327146.1 | 1 - 2 | 285 | 154 | |
| NM_001337856.1 | NP_001327145.1 | 1 - 2 | 285 | 154 | ||
| NM_001337857.1 | NP_001327144.1 | 1 - 2 | 285 | 154 | ||
| NM_111846.3 | NP_566368.1 | 1 - 2 | 285 | 154 | ||
| GenBank mRNA | BT030038.1 | - | 1 - 2 | 170 | 154 | |
More Information
Set Membership: |
|

MEE67
Genomic Map