Assay Details
| Gene Symbol: AT3G14390 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112297.3 | NP_188056.1 | 5 - 6 | 1264 | 142 | |
| GenBank mRNA | AF227913.1 | - | 5 - 6 | 1209 | 142 | |
| AY050823.1 | - | 5 - 6 | 1186 | 142 | ||
| AY088418.1 | - | 5 - 6 | 1196 | 142 | ||
| AY122998.1 | - | 5 - 6 | 1163 | 142 | ||
More Information
Set Membership: |
|

AT3G14390
Genomic Map