Assay Details
| Gene Symbol: AOX1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113134.2 | NP_188875.1 | 2 - 3 | 485 | 168 | |
| GenBank mRNA | DQ056603.1 | - | 2 - 3 | 426 | 168 | |
More Information
Set Membership: |
|

AOX1B
Genomic Map