Assay Details
| Gene Symbol: XRCC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161167.1 | NP_001154639.1 | 1 - 2 | 229 | 121 | |
| NM_113211.2 | NP_188951.2 | 1 - 2 | 229 | 121 | ||
| GenBank mRNA | AF233528.2 | - | 1 - 2 | 229 | 121 | |
More Information
Set Membership: |
|

XRCC4
Genomic Map