Assay Details
| Gene Symbol: AOX1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113678.3 | NP_189399.1 | 1 - 2 | 424 | 178 | |
| GenBank mRNA | BT004153.1 | - | 1 - 2 | 419 | 178 | |
| BT005467.1 | - | 1 - 2 | 312 | 178 | ||
More Information
Set Membership: |
|

AOX1C
Genomic Map