Assay Details
| Gene Symbol: AOX1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_113678.3 | NP_189399.1 | 3 - 4 | 1040 | 137 | |
| GenBank mRNA | BT004153.1 | - | 3 - 4 | 1035 | 137 | |
More Information
Set Membership: |
|

AOX1C
Genomic Map