Assay Details
Gene Symbol: IMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332948.1 | NP_001322636.1 | 1 - 2 | 446 | 105 | |
NM_102856.3 | NP_174403.1 | 1 - 2 | 379 | 105 | ||
GenBank mRNA | BT025974.1 | - | 1 - 2 | 346 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
