Assay Details
Gene Symbol: IMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332948.1 | NP_001322636.1 | 5 - 6 | 926 | 146 | |
NM_102856.3 | NP_174403.1 | 5 - 6 | 859 | 146 | ||
GenBank mRNA | BT025974.1 | - | 5 - 6 | 826 | 146 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
