Assay Details
| Gene Symbol: PIP2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035774.1 | NP_001030851.1 | 1 - 2 | 511 | 127 | |
| NM_115202.3 | NP_190910.1 | 1 - 2 | 578 | 127 | ||
| GenBank mRNA | AF428426.1 | - | 1 - 2 | 361 | 127 | |
| AY039579.1 | - | 1 - 2 | 361 | 127 | ||
| AY044327.1 | - | 1 - 2 | 302 | 127 | ||
| AY056085.1 | - | 1 - 2 | 302 | 127 | ||
| AY072374.1 | - | 1 - 2 | 353 | 127 | ||
| AY087854.1 | - | 1 - 2 | 563 | 127 | ||
| X75883.1 | - | 341 | 127 | |||
More Information
Set Membership: |
|

PIP2A
Genomic Map