Assay Details
| Gene Symbol: AUD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125421.1 | NP_001118893.1 | 4 - 5 | 973 | 162 | |
| NM_116148.3 | NP_191842.1 | 3 - 4 | 1113 | 162 | ||
| GenBank mRNA | AY143897.1 | - | 3 - 4 | 846 | 162 | |
More Information
Set Membership: |
|

AUD1
Genomic Map