Assay Details
| Gene Symbol: BRCA2(IV) | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160720.1 | NP_001154192.1 | 15 - 16 | 2611 | 96 | |
| NM_001340223.1 | NP_001328738.1 | 14 - 15 | 2766 | 96 | ||
| NM_001340224.1 | NP_001328737.1 | 14 - 15 | 2766 | 96 | ||
| NM_001340225.1 | NP_001328739.1 | 14 - 15 | 2766 | 96 | ||
| NM_116219.5 | NP_191913.3 | 14 - 15 | 2766 | 96 | ||
| GenBank mRNA | AJ488304.1 | - | 14 - 15 | 2626 | 96 | |
More Information
Set Membership: |
|

BRCA2(IV)
Genomic Map