Assay Details
| Gene Symbol: AT4G11320 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340705.1 | NP_001328659.1 | 3 - 4 | 983 | 144 | |
| NM_117203.4 | NP_567377.1 | 3 - 4 | 1044 | 144 | ||
| GenBank mRNA | AY035055.1 | - | 3 - 4 | 956 | 144 | |
| AY051062.1 | - | 3 - 4 | 874 | 144 | ||
More Information
Set Membership: |
|

AT4G11320
Genomic Map