Assay Details
| Gene Symbol: CRK6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118443.3 | NP_567678.1 | 1 - 2 | 869 | 133 | |
| NM_179095.1 | NP_849426.1 | 1 - 2 | 855 | 133 | ||
| GenBank mRNA | AF224706.1 | - | 1 - 2 | 841 | 133 | |
| AK118493.1 | - | 1 - 2 | 855 | 133 | ||
More Information
Set Membership: |
|

CRK6
Genomic Map