Assay Details
Gene Symbol: CRK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118443.3 | NP_567678.1 | 6 - 7 | 1722 | 122 | |
NM_179095.1 | NP_849426.1 | 6 - 7 | 1726 | 122 | ||
GenBank mRNA | AF224706.1 | - | 6 - 7 | 1694 | 122 | |
AK118493.1 | - | 6 - 7 | 1726 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
