Assay Details
| Gene Symbol: AT4G29630 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119108.1 | NP_194692.1 | 1 | 159 | 131 | |
| GenBank mRNA | DQ056662.1 | - | 1 | 159 | 131 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT4G29630| Gene Symbol: AT4G29630 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119108.1 | NP_194692.1 | 1 | 159 | 131 | |
| GenBank mRNA | DQ056662.1 | - | 1 | 159 | 131 | |
Set Membership: |
|