Assay Details
| Gene Symbol: RAC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119762.4 | NP_195320.1 | 2 - 3 | 537 | 148 | |
| GenBank mRNA | AF079487.1 | - | 2 - 3 | 417 | 148 | |
| AF107663.1 | - | 2 - 3 | 244 | 148 | ||
| AK117209.1 | - | 2 - 3 | 534 | 148 | ||
| BT005217.1 | - | 2 - 3 | 185 | 148 | ||
More Information
Set Membership: |
|

RAC6
Genomic Map