Assay Details
| Gene Symbol: AT5G03810 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120462.2 | NP_196001.2 | 2 - 3 | 372 | 142 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT5G03810| Gene Symbol: AT5G03810 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120462.2 | NP_196001.2 | 2 - 3 | 372 | 142 | |
Set Membership: |
|