Assay Details
| Gene Symbol: NIP4;1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123140.2 | NP_198597.1 | 1 - 2 | 251 | 114 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NIP4;1| Gene Symbol: NIP4;1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123140.2 | NP_198597.1 | 1 - 2 | 251 | 114 | |
Set Membership: |
|