Assay Details
Gene Symbol: AT5G38030 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123162.4 | NP_198619.1 | 2 - 3 | 1103 | 169 | |
GenBank mRNA | AF372972.1 | - | 2 - 3 | 906 | 169 | |
BT002707.1 | - | 2 - 3 | 839 | 169 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
