Assay Details
| Gene Symbol: AGL77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123235.2 | NP_198690.1 | 1 | 1040 | 87 | |
| GenBank mRNA | AB493770.1 | - | 1 | 1040 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AGL77| Gene Symbol: AGL77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123235.2 | NP_198690.1 | 1 | 1040 | 87 | |
| GenBank mRNA | AB493770.1 | - | 1 | 1040 | 87 | |
Set Membership: |
|