Assay Details
Gene Symbol: RHA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344637.1 | NP_001331800.1 | 1 - 2 | 932 | 100 | |
NM_123938.4 | NP_199383.1 | 2 - 3 | 726 | 100 | ||
GenBank mRNA | AK229058.1 | - | 2 - 3 | 539 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
