Assay Details
| Gene Symbol: TUB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125665.4 | NP_568960.1 | 2 - 3 | 907 | 171 | |
| Gene Symbol: TUB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY060550.1 | - | 2 - 3 | 766 | 171 | |
More Information
Set Membership: |
|

TUB3
Genomic Map