Assay Details
| Gene Symbol: PAD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126012.4 | NP_201415.1 | 1 - 2 | 266 | 148 | |
| GenBank mRNA | AF043523.1 | - | 1 - 2 | 212 | 148 | |
More Information
Set Membership: |
|

PAD2
Genomic Map