Assay Details
Gene Symbol: ERF1-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101163.2 | NP_172752.1 | 2 | 1498 | 135 | |
GenBank mRNA | AY034963.1 | - | 2 | 1451 | 135 | |
AY086623.1 | - | 2 | 1455 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
