Assay Details
Gene Symbol: HB31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101311.6 | NP_172896.1 | 2 | 1917 | 113 | |
GenBank mRNA | AY074266.1 | - | 2 | 1266 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
