Assay Details
Gene Symbol: PUT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102920.3 | NP_174466.1 | 2 | 1283 | 97 | |
NM_202218.3 | NP_973947.1 | 2 | 1216 | 97 | ||
GenBank mRNA | AK317082.1 | - | 2 | 1071 | 97 | |
BT030375.1 | - | 1 | 944 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
