Assay Details
| Gene Symbol: CAM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339108.1 | NP_001326524.1 | 2 | 1845 | 79 | |
| NM_001339109.1 | NP_001326522.1 | 2 | 985 | 79 | ||
| NM_114249.4 | NP_189967.1 | 2 | 739 | 79 | ||
More Information
Set Membership: |
|

CAM7
Genomic Map